This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02408175_g1
See other GLD-2 GE Assays ›
Gene Symbol:
gld-2
Gene Name:
protein-coding
Gene Aliases:
CELE_ZC308.1
Chromosome Location:
Chr.I: 6369748 - 6386194 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02408175_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: gld-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001026673.3 NP_001021844.1 3 - 4 1086 105
NM_001313548.3 NP_001300477.1 7 - 8 955 105
NM_001313549.3 NP_001300478.1 5 - 6 676 105
NM_059440.5 NP_491841.2 6 - 7 1559 105
NM_059441.5 NP_491842.2 14 - 15 1969 105
GenBank mRNA AY125085.1 - 13 - 14 1817 105

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mRNA polyadenylation
mRNA processing
nucleus organization
multicellular organism development
embryo development ending in birth or egg hatching
RNA polyadenylation
regulation of cell division
meiotic cell cycle
positive regulation of meiosis I
nucleotide binding
polynucleotide adenylyltransferase activity
protein binding
ATP binding
transferase activity
nucleotidyltransferase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0