Assay Details
Gene Symbol: C48E7.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001374899.2 | NP_001361945.1 | 17 - 18 | 3385 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: C48E7.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001374899.2 | NP_001361945.1 | 17 - 18 | 3385 | 147 |
Set Membership: |
![]() ![]() ![]() ![]() |