This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02407973_g1
See other FARS-1 GE Assays ›
Gene Symbol:
fars-1 mrpl-23
Gene Name:
protein-coding
Probable 39S ribosomal protein L23, mitochondrial
Gene Aliases:
CELE_T08B2.9
CELE_T08B2.8
Chromosome Location:
Chr.I: 6211981 - 6214645 on Build WBcel235
Chromosome Location:
Chr.I: 6211266 - 6214586 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02407973_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fars-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_059391.5 NP_491792.1 1 - 2 63 120
Gene Symbol: mrpl-23
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001380479.1 NP_001367676.1 1 - 2 64 120

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001380479.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
translation
phenylalanyl-tRNA aminoacylation
tRNA aminoacylation
mitochondrial translation
tRNA binding
nucleotide binding
aminoacyl-tRNA ligase activity
phenylalanine-tRNA ligase activity
ATP binding
ligase activity
structural constituent of ribosome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0