This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02407486_g1
See other UNC-77 GE Assays ›
Gene Symbol:
unc-77
Gene Name:
protein-coding
Gene Aliases:
CELE_C11D2.6
Chromosome Location:
Chr.IV: 6157306 - 6183857 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02407486_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: unc-77
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001306889.3 NP_001293818.1 25 - 26 5212 82
NM_001306890.3 NP_001293819.1 24 - 25 5032 82
NM_001306891.3 NP_001293820.1 25 - 26 5176 82
NM_001306892.3 NP_001293821.1 24 - 25 5074 82
NM_001306893.3 NP_001293822.1 24 - 25 5074 82
NM_001306894.3 NP_001293823.1 25 - 26 5176 82
NM_001306895.3 NP_001293824.1 26 - 27 5230 82
NM_001306896.3 NP_001293825.1 25 - 26 5128 82
NM_001306897.3 NP_001293826.1 25 - 26 5128 82
NM_001306898.3 NP_001293827.1 26 - 27 5230 82
NM_001307702.3 NP_001294631.1 24 - 25 5110 82
NM_001307703.3 NP_001294632.1 23 - 24 4930 82
NM_068525.4 NP_500926.3 24 - 25 5213 82
NM_068526.5 NP_500927.3 23 - 24 4930 82
NM_171352.5 NP_741413.2 25 - 26 5212 82
NM_171353.6 NP_741414.2 24 - 25 5032 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ion transport
positive regulation of synaptic transmission, cholinergic
positive regulation of synaptic transmission, GABAergic
ion transmembrane transport
positive regulation of locomotion
transmembrane transport
cation transmembrane transport
ion channel activity
cation channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0