This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02407480_m1
See other UNC-77 GE Assays ›
Gene Symbol:
unc-77
Gene Name:
protein-coding
Gene Aliases:
CELE_C11D2.6
Chromosome Location:
Chr.IV: 6157306 - 6183857 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02407480_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: unc-77
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001306889.3 NP_001293818.1 19 - 20 3312 77
NM_001306890.3 NP_001293819.1 18 - 19 3132 77
NM_001306891.3 NP_001293820.1 19 - 20 3276 77
NM_001306892.3 NP_001293821.1 18 - 19 3174 77
NM_001306893.3 NP_001293822.1 18 - 19 3174 77
NM_001306894.3 NP_001293823.1 19 - 20 3276 77
NM_001306895.3 NP_001293824.1 20 - 21 3330 77
NM_001306896.3 NP_001293825.1 19 - 20 3228 77
NM_001306897.3 NP_001293826.1 19 - 20 3228 77
NM_001306898.3 NP_001293827.1 20 - 21 3330 77
NM_001307702.3 NP_001294631.1 18 - 19 3210 77
NM_001307703.3 NP_001294632.1 17 - 18 3030 77
NM_068525.4 NP_500926.3 18 - 19 3313 77
NM_068526.5 NP_500927.3 17 - 18 3030 77
NM_171352.5 NP_741413.2 19 - 20 3312 77
NM_171353.6 NP_741414.2 18 - 19 3132 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ion transport
positive regulation of synaptic transmission, cholinergic
positive regulation of synaptic transmission, GABAergic
ion transmembrane transport
positive regulation of locomotion
transmembrane transport
cation transmembrane transport
ion channel activity
cation channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0