This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02407469_g1
See other UNC-77 GE Assays ›
Gene Symbol:
unc-77
Gene Name:
protein-coding
Gene Aliases:
CELE_C11D2.6
Chromosome Location:
Chr.IV: 6157306 - 6183857 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02407469_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: unc-77
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001306889.3 NP_001293818.1 9 - 10 1772 92
NM_001306890.3 NP_001293819.1 8 - 9 1592 92
NM_001306891.3 NP_001293820.1 9 - 10 1736 92
NM_001306892.3 NP_001293821.1 9 - 10 1736 92
NM_001306893.3 NP_001293822.1 9 - 10 1736 92
NM_001306894.3 NP_001293823.1 9 - 10 1736 92
NM_001306895.3 NP_001293824.1 10 - 11 1790 92
NM_001306896.3 NP_001293825.1 10 - 11 1790 92
NM_001306897.3 NP_001293826.1 10 - 11 1790 92
NM_001306898.3 NP_001293827.1 10 - 11 1790 92
NM_001307702.3 NP_001294631.1 9 - 10 1772 92
NM_001307703.3 NP_001294632.1 8 - 9 1592 92
NM_068525.4 NP_500926.3 9 - 10 1875 92
NM_068526.5 NP_500927.3 8 - 9 1592 92
NM_171352.5 NP_741413.2 9 - 10 1772 92
NM_171353.6 NP_741414.2 8 - 9 1592 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ion transport
positive regulation of synaptic transmission, cholinergic
positive regulation of synaptic transmission, GABAergic
ion transmembrane transport
positive regulation of locomotion
transmembrane transport
cation transmembrane transport
ion channel activity
cation channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0