This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02407363_g1
See other YARS-2 GE Assays ›
Gene Symbol:
yars-2
Gene Name:
protein-coding
Gene Aliases:
CELE_K08F11.4
Chromosome Location:
Chr.IV: 4710861 - 4712553 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02407363_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: yars-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001383084.1 2 - 3 214 70
NM_171324.5 NP_741382.1 2 - 3 173 70

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001383084.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

translation
tRNA aminoacylation for protein translation
tyrosyl-tRNA aminoacylation
tRNA aminoacylation
nucleotide binding
aminoacyl-tRNA ligase activity
tyrosine-tRNA ligase activity
ATP binding
ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0