This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02406971_g1
See other HENN-1 GE Assays ›
Gene Symbol:
henn-1
Gene Name:
Small RNA 2'-O-methyltransferase
Gene Aliases:
CELE_C02F5.6
Chromosome Location:
Chr.III: 8232651 - 8234614 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02406971_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: henn-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001322688.2 NP_001309472.1 5 - 6 1104 110
NR_136348.1 5 - 6 1097 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

RNA methylation
production of siRNA involved in RNA interference
gene silencing by RNA
methylation
piRNA metabolic process
regulation of production of siRNA involved in RNA interference
RNA binding
methyltransferase activity
O-methyltransferase activity
RNA methyltransferase activity
transferase activity
metal ion binding
small RNA 2'-O-methyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0