This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02406326_g1
See other PAT-12 GE Assays ›
Gene Symbol:
pat-12
Gene Name:
protein-coding
Gene Aliases:
CELE_T17H7.4
Chromosome Location:
Chr.III: 727207 - 752432 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02406326_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pat-12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001373834.2 NP_001359886.1 11 - 12 1139 110
NM_001381698.1 NP_001367550.1 9 - 10 949 110
NM_001381699.1 NP_001367549.1 11 - 12 1133 110
NM_001381700.1 NP_001367551.1 12 - 13 1258 110
NM_001381701.1 NP_001367554.1 3 - 4 211 110
NM_001381702.1 NP_001367555.1 4 - 5 340 110
NM_001381703.2 NP_001367552.1 8 - 9 2223 110
NM_001381704.2 NP_001367553.1 13 - 14 1320 110
NM_001384009.1 14 - 15 3829 110
NM_171067.5 NP_741076.1 13 - 14 1314 110
NM_171068.5 NP_741077.1 11 - 12 1140 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

hemidesmosome assembly
embryonic morphogenesis
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0