This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02406197_g1
See other PTP-3 GE Assays ›
Gene Symbol:
ptp-3
Gene Name:
protein-coding
Gene Aliases:
CELE_C09D8.1
Chromosome Location:
Chr.II: 10975539 - 11012254 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02406197_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ptp-3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001026771.3 NP_001021942.1 11 - 12 1811 83
NM_001267341.4 NP_001254270.1 8 - 9 1266 83
NM_001276741.3 NP_001263670.1 12 - 13 1883 83
NM_001276742.3 NP_001263671.1 12 - 13 2015 83
NM_001276743.3 NP_001263672.1 11 - 12 1943 83
NM_001377858.3 NP_001364824.1 12 - 13 1883 83
NM_001377859.2 NP_001364823.1 11 - 12 1811 83
NM_001377860.2 NP_001364658.1 9 - 10 1298 83
NM_001377861.2 NP_001364506.1 9 - 10 1336 83
NM_001377862.2 NP_001364507.1 8 - 9 1226 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
protein dephosphorylation
gastrulation
nervous system development
synapse assembly
motor neuron axon guidance
epidermis development
embryonic body morphogenesis
peptidyl-tyrosine dephosphorylation
protein localization to synapse
neuroblast migration
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
transmembrane receptor protein tyrosine phosphatase activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0