This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02405696_m1
See other HMR-1 GE Assays ›
Gene Symbol:
hmr-1
Gene Name:
protein-coding
Gene Aliases:
CELE_W02B9.1
Chromosome Location:
Chr.I: 11075755 - 11134667 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02405696_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: hmr-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001026479.4 NP_001021650.1 27 - 28 7058 79
NM_001264632.3 NP_001251561.1 4 - 5 1382 79
NM_001381215.1 NP_001366962.1 7 - 8 2223 79
NM_001381216.1 NP_001368105.1 2 - 3 992 79
GenBank mRNA AJ307058.1 - 27 - 28 7058 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

establishment of mitotic spindle orientation
apical constriction involved in gastrulation
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
multicellular organism development
gastrulation
nervous system development
embryo development ending in birth or egg hatching
cell migration
cell differentiation
cortical actin cytoskeleton organization
cell migration involved in gastrulation
cell-cell adhesion
calcium ion binding
protein binding
beta-catenin binding
delta-catenin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0