This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02405396_g1
See other F46F11.1 GE Assays ›
Gene Symbol:
F46F11.1
Gene Name:
protein-coding
Gene Aliases:
CELE_F46F11.1
Chromosome Location:
Chr.I: 5627813 - 5637622 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02405396_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: F46F11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_170868.5 NP_740855.2 3 - 4 258 135
NM_171837.9 NP_740856.1 3 - 4 264 135

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

inositol metabolic process
phosphorylation
inositol phosphate biosynthetic process
nucleotide binding
inositol-1,3,4,5,6-pentakisphosphate kinase activity
inositol hexakisphosphate kinase activity
inositol heptakisphosphate kinase activity
inositol hexakisphosphate 5-kinase activity
ATP binding
kinase activity
transferase activity
diphosphoinositol-pentakisphosphate kinase activity
inositol hexakisphosphate 1-kinase activity
inositol hexakisphosphate 3-kinase activity
5-diphosphoinositol pentakisphosphate 3-kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0