This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02405271_m1
See other UBR-4 GE Assays ›
Gene Symbol:
ubr-4
Gene Name:
protein-coding
Gene Aliases:
CELE_C44E4.1
Chromosome Location:
Chr.I: 4610143 - 4633379 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02405271_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ubr-4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318271.2 NP_001305200.1 14 - 15 8736 73
NM_001382970.1 14 - 15 8765 73
NM_059006.3 NP_491407.2 12 - 13 8715 73
NR_133620.1 11 - 12 9472 73

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001382970.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0