This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02405079_g1
See other C53H9.2 GE Assays ›
Gene Symbol:
C53H9.2 rpl-27
Gene Name:
CP-type G domain-containing protein
60S ribosomal protein L27
Gene Aliases:
CELE_C53H9.2
CELE_C53H9.1
Chromosome Location:
Chr.I: 1832897 - 1834763 on Build WBcel235
Chromosome Location:
Chr.I: 1832890 - 1836655 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02405079_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C53H9.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_170804.6 NP_740787.2 1 - 2 278 71
NR_131348.1 1 - 2 278 71
NR_131349.1 1 - 2 278 71
NR_131350.1 1 - 2 278 71
NR_151555.1 1 - 2 278 71
Gene Symbol: rpl-27
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001381283.2 NP_001367687.1 1 - 2 285 71

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001381283.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ribosomal subunit export from nucleus
translation
GTPase activity
GTP binding
structural constituent of ribosome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0