This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02403154_g1
See other SNT-7 GE Assays ›
Gene Symbol:
snt-7
Gene Name:
C2 domain-containing protein
Gene Aliases:
CELE_T10B10.5
Chromosome Location:
Chr.X: 15195883 - 15199152 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02403154_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: snt-7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_078125.6 NP_510526.2 7 - 8 861 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of dopamine secretion
synaptic vesicle exocytosis
vesicle-mediated transport
calcium ion regulated exocytosis
regulation of calcium ion-dependent exocytosis
synaptic vesicle endocytosis
calcium ion-regulated exocytosis of neurotransmitter
cellular response to calcium ion
SNARE binding
phosphatidylserine binding
calcium ion binding
calcium-dependent phospholipid binding
syntaxin binding
clathrin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0