This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02402466_g1
See other TAP-1 GE Assays ›
Gene Symbol:
tap-1
Gene Name:
TGF-beta-activated kinase 1 and MAP3K7-binding protein 1
Gene Aliases:
CELE_C44H4.5
Chromosome Location:
Chr.X: 14587505 - 14591823 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02402466_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: tap-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_078027.6 NP_510428.1 5 - 6 706 99
GenBank mRNA AF145375.1 - 4 - 5 500 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

endodermal cell fate specification
protein dephosphorylation
activation of protein kinase activity
negative regulation of stress-activated MAPK cascade
positive regulation of protein kinase activity
negative regulation of Ras protein signal transduction
catalytic activity
protein serine/threonine phosphatase activity
protein kinase activator activity
mitogen-activated protein kinase kinase kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0