Assay Details
Gene Symbol: N4BP2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC120192.1 | - | 1 - 2 | 108 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

Gene Symbol: N4BP2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC120192.1 | - | 1 - 2 | 108 | 88 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |