This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt07107831_s1
See other PTGDR2 GE Assays ›
Gene Symbol:
PTGDR2
Gene Name:
prostaglandin D2 receptor 2
Gene Aliases:
CRTH2, GPR44
Chromosome Location:
Chr.29: 37178760 - 37187630 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt07107831_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTGDR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AY194233.1 - 1 222 133

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm