Assay Details
Gene Symbol: GCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC148907.1 | - | 1 - 2 | 132 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Gene Symbol: GCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC148907.1 | - | 1 - 2 | 132 | 80 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |