This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04647062_s1
See other MIR320A-1 GE Assays ›
Gene Symbol:
MIR320A-1
Gene Name:
microRNA mir-320a-1
Gene Aliases:
MIR320-1, bta-mir-320-1, bta-mir-320a-1, mir-320a-1
Chromosome Location:
Chr.8: 69558096 - 69558177 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04647062_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR320A-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_036340.1 1 45 73

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream phytanoyl-CoA 2-hydroxylase interacting protein PHYHIP -13.8kb Sense Chr. 8 - 69531549 - 69544277 on NCBI Build ARS-UCD1.2
Downstream RNA polymerase III subunit D POLR3D 26.0 Anti-sense Chr. 8 - 69558203 - 69564549 on NCBI Build ARS-UCD1.2

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Bos taurus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0