This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04647060_s1
See other MIR214 GE Assays ›
Gene Symbol:
MIR214
Gene Name:
microRNA mir-214
Gene Aliases:
MIRN214, bta-mir-214
Chromosome Location:
Chr.16: 39619549 - 39619658 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04647060_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR214
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_031345.1 1 53 104

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA mir-3120 MIR3120 bta-mir-3120 Anti-sense Chr. 16 - 39619559 - 39619639 on NCBI Build ARS-UCD1.2
Downstream microRNA mir-199a-1 MIR199A-1 MIRN199A,
bta-mir-199a,
bta-mir-199a-1,
mir-199a-1
5.7kb Sense Chr. 16 - 39625344 - 39625447 on NCBI Build ARS-UCD1.2

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Bos taurus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0