This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04646800_s1
See other RMRP GE Assays ›
Gene Symbol:
RMRP
Gene Name:
RNA component of mitochondrial RNA processing endoribonuclease
Gene Aliases:
-
Chromosome Location:
Chr.8: 59833115 - 59833395 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04646800_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RMRP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_036646.1 1 124 90

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream signaling threshold regulating transmembrane adaptor 1 SIT1 -4.0kb Sense Chr. 8 - 59827540 - 59829096 on NCBI Build ARS-UCD1.2
Downstream coiled-coil domain containing 107 CCDC107 303.0 Anti-sense Chr. 8 - 59833698 - 59836258 on NCBI Build ARS-UCD1.2

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Bos taurus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0