This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04646706_s1
See other MIR320A-2 GE Assays ›
Gene Symbol:
MIR320A-2
Gene Name:
microRNA mir-320a-2
Gene Aliases:
MIR320, MIR320A, MIRN320, MIRN320A, bta-mir-320, bta-mir-320a-2, mir-320a-2
Chromosome Location:
Chr.20: 15245796 - 15245877 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04646706_s1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR320A-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030924.1 1 42 67

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream regulator of G protein signaling 7 binding protein RGS7BP -272.3kb Anti-sense Chr. 20 - 14840630 - 14973460 on NCBI Build ARS-UCD1.2
Downstream 5-hydroxytryptamine receptor 1A HTR1A 5HT1A,
5htr1a
341.2kb Sense Chr. 20 - 15587097 - 15589998 on NCBI Build ARS-UCD1.2

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0