This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04646670_s1
See other MIR493 GE Assays ›
Gene Symbol:
MIR493
Gene Name:
microRNA mir-493
Gene Aliases:
bta-mir-493
Chromosome Location:
Chr.21: 65766680 - 65766768 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04646670_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR493
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_031330.1 1 45 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream maternally expressed 3 (non-protein coding) MEG3 Glt2,
Gtl2
-20.6kb Sense Chr. 21 - 65727804 - 65746033 on NCBI Build ARS-UCD1.2
Downstream microRNA mir-665 MIR665 bta-mir-665 7.6kb Sense Chr. 21 - 65774396 - 65774467 on NCBI Build ARS-UCD1.2

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0