This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04418217_s1
See other MIR423 GE Assays ›
Gene Symbol:
MIR423
Gene Name:
microRNA mir-423
Gene Aliases:
MIRN423, bta-mir-423, mir-423
Chromosome Location:
Chr.19: 21238007 - 21238100 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04418217_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR423
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_030961.1 1 50 68

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream coronin 6 CORO6 -364.4kb Anti-sense Chr. 19 - 20865343 - 20873589 on NCBI Build ARS-UCD1.2
Downstream solute carrier family 6 member 4 SLC6A4 SERT 52.3kb Anti-sense Chr. 19 - 21290406 - 21323579 on NCBI Build ARS-UCD1.2

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0