This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt04286161_g1
See other ADGRA1 GE Assays ›
Gene Symbol:
ADGRA1
Gene Name:
adhesion G protein-coupled receptor A1
Gene Aliases:
GPR123
Chromosome Location:
Chr.26: 50687922 - 50723787 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt04286161_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001205846.1 NP_001192775.1 1 - 2 144 75

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001205846.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0