This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03287790_s1
See other SFN GE Assays ›
Gene Symbol:
SFN
Gene Name:
stratifin
Gene Aliases:
-
Chromosome Location:
Chr.2: 126340193 - 126341536 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03287790_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SFN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001075912.1 NP_001069380.1 1 1248 140
GenBank mRNA BC120373.1 - 1 1248 140

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of cyclin-dependent protein serine/threonine kinase activity
release of cytochrome c from mitochondria
keratinocyte development
intrinsic apoptotic signaling pathway in response to DNA damage
regulation of epidermal cell division
negative regulation of keratinocyte proliferation
positive regulation of cell growth
keratinization
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of epidermal cell differentiation
positive regulation of protein export from nucleus
establishment of skin barrier
protein kinase binding
identical protein binding
phosphoprotein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0