This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03276634_m1
See other CAMK2N1 GE Assays ›
Gene Symbol:
CAMK2N1
Gene Name:
calcium/calmodulin dependent protein kinase II inhibitor 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 132136807 - 132139924 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03276634_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CAMK2N1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114520.1 NP_001107992.1 1 - 2 228 68
GenBank mRNA BC151544.1 - 1 - 2 228 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

kinase inhibitor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cell proliferation
positive regulation of gene expression
negative regulation of cell cycle
negative regulation of proteolysis
negative regulation of ERK1 and ERK2 cascade
negative regulation of cyclin-dependent protein kinase activity
calcium-dependent protein kinase inhibitor activity
protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0