Assay Details
| Gene Symbol: GPR62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101238.2 | NP_001094708.1 | 1 - 2 | 128 | 71 | |
| GenBank mRNA | BC151553.1 | - | 1 - 2 | 129 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC151553.1, NM_001101238.2 transcripts.
Set Membership: |
|

GPR62
Genomic Map