This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03269627_m1
See other PLAAT1 GE Assays ›
Gene Symbol:
PLAAT1
Gene Name:
phospholipase A and acyltransferase 1
Gene Aliases:
HRASLS
Chromosome Location:
Chr.1: 74163663 - 74183483 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03269627_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLAAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001076225.3 NP_001069693.2 3 - 4 549 69
NM_001314065.1 NP_001300994.1 3 - 4 248 69
GenBank mRNA BC114073.1 - 3 - 4 543 69

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC114073.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0