This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03259262_m1
See other ACVR2A GE Assays ›
Gene Symbol:
ACVR2A
Gene Name:
activin A receptor type 2A
Gene Aliases:
ACVR2
Chromosome Location:
Chr.2: 48279645 - 48374634 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03259262_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACVR2A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174227.3 NP_776652.1 9 - 10 1770 67
GenBank mRNA BC140605.1 - 9 - 10 1770 67
L21717.1 - 9 - 10 1238 67
U43208.1 - 9 - 10 1219 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

serine/threonine protein kinase receptor

Gene Ontology Categories:

Function(s) Process(es)

gastrulation with mouth forming second
spermatogenesis
determination of left/right symmetry
mesoderm development
anterior/posterior pattern specification
positive regulation of pathway-restricted SMAD protein phosphorylation
positive regulation of bone mineralization
BMP signaling pathway
positive regulation of activin receptor signaling pathway
sperm ejaculation
penile erection
positive regulation of erythrocyte differentiation
positive regulation of osteoblast differentiation
positive regulation of transcription from RNA polymerase II promoter
embryonic skeletal system development
regulation of nitric-oxide synthase activity
Sertoli cell proliferation
ATP binding
coreceptor activity
growth factor binding
PDZ domain binding
inhibin binding
protein self-association
metal ion binding
BMP receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0