This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03257329_m1
See other NOL3 GE Assays ›
Gene Symbol:
NOL3
Gene Name:
nucleolar protein 3
Gene Aliases:
-
Chromosome Location:
Chr.18: 34808439 - 34812047 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03257329_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NOL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001083688.2 NP_001077157.1 1 - 2 154 68
GenBank mRNA BC134717.1 - 1 - 2 155 68

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC134717.1, NM_001083688.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0