This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03255013_m1
See other IFITM5 GE Assays ›
Gene Symbol:
IFITM5
Gene Name:
interferon induced transmembrane protein 5
Gene Aliases:
-
Chromosome Location:
Chr.11: 82964858 - 82966228 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03255013_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IFITM5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081724.2 NP_001075193.1 1 - 2 228 55
GenBank mRNA BC126775.1 - 1 - 2 229 55

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0