Assay Details
Gene Symbol: SCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080243.1 | NP_001073712.1 | 5 - 6 | 800 | 106 | |
GenBank mRNA | BC126613.1 | - | 5 - 6 | 800 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
