This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03249353_m1
See other TMBIM6 GE Assays ›
Gene Symbol:
TMBIM6
Gene Name:
transmembrane BAX inhibitor motif containing 6
Gene Aliases:
TEGT
Chromosome Location:
Chr.5: 30112133 - 30128722 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03249353_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMBIM6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001076414.1 NP_001069882.1 9 - 10 760 102
GenBank mRNA BC102469.1 - 9 - 10 774 102
BC149995.1 - 9 - 10 823 102
BT026337.1 - 9 - 10 760 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

autophagy
negative regulation of calcium ion transport into cytosol
negative regulation of protein binding
endoplasmic reticulum calcium ion homeostasis
negative regulation of RNA splicing
cellular response to unfolded protein
negative regulation of immunoglobulin secretion
negative regulation of endoribonuclease activity
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
response to L-glutamate
negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
negative regulation of hypoxia-induced intrinsic apoptotic signaling pathway
negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
ubiquitin protein ligase binding
endoribonuclease inhibitor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0