This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03241943_m1
See other GPRC5B GE Assays ›
Gene Symbol:
GPRC5B
Gene Name:
G protein-coupled receptor class C group 5 member B
Gene Aliases:
-
Chromosome Location:
Chr.25: 17477366 - 17498791 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03241943_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPRC5B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001075741.1 NP_001069209.1 1 - 2 108 95
GenBank mRNA BC149136.1 - 1 - 2 92 95
BT025414.1 - 1 - 2 108 95

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC149136.1, NM_001075741.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0