This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03241410_m1
See other ALDH3B1 GE Assays ›
Gene Symbol:
ALDH3B1
Gene Name:
aldehyde dehydrogenase 3 family member B1
Gene Aliases:
-
Chromosome Location:
Chr.29: 45537445 - 45557502 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03241410_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDH3B1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001075518.1 NP_001068986.1 1 - 2 643 115
GenBank mRNA BC147958.1 - 2 - 3 189 115
BT025453.1 - 1 - 2 643 115
BT026328.1 - 2 - 3 443 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

ethanol catabolic process
cellular aldehyde metabolic process
oxidation-reduction process
benzaldehyde dehydrogenase (NADP+) activity
benzaldehyde dehydrogenase (NAD+) activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0