This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03240039_m1
See other VAMP5 GE Assays ›
Gene Symbol:
VAMP5
Gene Name:
vesicle associated membrane protein 5
Gene Aliases:
-
Chromosome Location:
Chr.11: 49416822 - 49425368 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03240039_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VAMP5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001046476.1 NP_001039941.1 1 - 2 46 112
GenBank mRNA BC112840.1 - 1 - 2 46 112

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC112840.1, NM_001046476.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons

Gene Ontology Categories:

Function(s)

muscle organ development
cell differentiation

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0