This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03239612_g1
See other TENT2 GE Assays ›
Gene Symbol:
TENT2
Gene Name:
terminal nucleotidyltransferase 2
Gene Aliases:
PAPD4
Chromosome Location:
Chr.10: 10611606 - 10672728 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03239612_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TENT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001046361.1 NP_001039826.1 1 - 2 309 82
GenBank mRNA BC113319.1 - 1 - 2 309 82

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC113319.1, NM_001046361.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

RNA stabilization
RNA polyadenylation
histone mRNA catabolic process
negative regulation of miRNA catabolic process
polynucleotide adenylyltransferase activity
ATP binding
RNA polymerase activity
metal ion binding
adenylyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0