This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03236935_m1
See other TMBIM7 GE Assays ›
Gene Symbol:
TMBIM7
Gene Name:
transmembrane BAX inhibitor motif containing 7
Gene Aliases:
TMBIM1B
Chromosome Location:
Chr.4: 9735082 - 9759260 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03236935_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMBIM7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001077068.1 NP_001070536.1 3 - 4 433 83
GenBank mRNA BC110220.1 - 3 - 4 433 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0