This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03235260_m1
See other STING1 GE Assays ›
Gene Symbol:
STING1
Gene Name:
stimulator of interferon response cGAMP interactor 1
Gene Aliases:
STING, TMEM173
Chromosome Location:
Chr.7: 50732084 - 50743011 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03235260_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STING1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001046357.2 NP_001039822.1 2 - 3 148 66
GenBank mRNA BC112716.1 - 2 - 3 149 66
KU695665.1 - 1 - 2 77 66
KU695666.1 - 1 - 2 77 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC112716.1, NM_001046357.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

autophagosome assembly
activation of innate immune response
positive regulation of defense response to virus by host
positive regulation of macroautophagy
positive regulation of protein binding
positive regulation of type I interferon production
interferon-beta production
cellular response to interferon-beta
innate immune response
positive regulation of transcription from RNA polymerase II promoter
regulation of inflammatory response
positive regulation of sequence-specific DNA binding transcription factor activity
protein oligomerization
defense response to virus
reticulophagy
cellular response to exogenous dsRNA
transcription factor binding
protein kinase binding
ubiquitin protein ligase binding
cyclic-di-GMP binding
protein homodimerization activity
cyclic-GMP-AMP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm