This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03235036_m1
See other MINDY2 GE Assays ›
Gene Symbol:
MINDY2
Gene Name:
MINDY lysine 48 deubiquitinase 2
Gene Aliases:
FAM63B
Chromosome Location:
Chr.10: 51423138 - 51508913 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03235036_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MINDY2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001046275.2 NP_001039740.1 6 - 7 1450 113
GenBank mRNA BC112797.1 - 6 - 7 1451 113

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons

Gene Ontology Categories:

Process(es)

thiol-dependent ubiquitin-specific protease activity
K48-linked polyubiquitin binding
K63-linked polyubiquitin binding
K11-linked polyubiquitin modification-dependent protein binding
K6-linked polyubiquitin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0