This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03230770_g1
See other S100A14 GE Assays ›
Gene Symbol:
S100A14
Gene Name:
S100 calcium binding protein A14
Gene Aliases:
-
Chromosome Location:
Chr.3: 16766172 - 16768623 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03230770_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100A14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001079634.2 NP_001073102.1 2 - 3 135 69
GenBank mRNA BC105161.1 - 2 - 3 136 69

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC105161.1, NM_001079634.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
response to lipopolysaccharide
toll-like receptor 4 signaling pathway
defense response to bacterium
positive regulation of granulocyte chemotaxis
positive regulation of monocyte chemotaxis
calcium ion binding
chemokine receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0