Assay Details
Gene Symbol: CD59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037446.2 | NP_001032523.1 | 1 - 2 | 45 | 53 | |
GenBank mRNA | BC108200.1 | - | 1 - 2 | 46 | 53 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC108200.1, NM_001037446.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
