This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03228772_g1
See other BPGM GE Assays ›
Gene Symbol:
BPGM
Gene Name:
bisphosphoglycerate mutase
Gene Aliases:
-
Chromosome Location:
Chr.4: 98321779 - 98353161 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03228772_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BPGM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001035402.2 NP_001030479.1 1 - 2 114 61
GenBank mRNA BC102611.1 - 1 - 2 115 61

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC102611.1, NM_001035402.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

mutase

Gene Ontology Categories:

Function(s) Process(es)

glycolytic process
bisphosphoglycerate mutase activity
phosphoglycerate mutase activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0