This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03228548_m1
See other ACAA2 GE Assays ›
Gene Symbol:
ACAA2
Gene Name:
acetyl-CoA acyltransferase 2
Gene Aliases:
-
Chromosome Location:
Chr.24: 49442437 - 49477387 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03228548_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACAA2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001035342.2 NP_001030419.1 9 - 10 1158 75
GenBank mRNA BC102287.1 - 9 - 10 1159 75
BT025480.1 - 9 - 10 1148 75
EF583004.1 - 9 - 10 1166 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

fatty acid beta-oxidation
negative regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
acetyl-CoA C-acetyltransferase activity
acetyl-CoA hydrolase activity
acetyl-CoA C-acyltransferase activity
palmitoyl-CoA hydrolase activity
acyl-CoA hydrolase activity
myristoyl-CoA hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0