This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03228115_m1
See other WRAP53 GE Assays ›
Gene Symbol:
WRAP53
Gene Name:
WD repeat containing antisense to TP53
Gene Aliases:
WDR79
Chromosome Location:
Chr.19: 27389505 - 27401320 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03228115_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: WRAP53
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001035073.1 NP_001030245.1 6 - 7 1067 76
GenBank mRNA BC104583.1 - 6 - 7 1067 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
telomere maintenance via telomerase
Cajal body organization
telomere formation via telomerase
RNA folding
positive regulation of DNA repair
positive regulation of telomerase activity
scaRNA localization to Cajal body
protein localization to Cajal body
positive regulation of double-strand break repair via homologous recombination
positive regulation of double-strand break repair
positive regulation of double-strand break repair via nonhomologous end joining
RNA binding
telomerase RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0