This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03226775_m1
See other SNAI2 GE Assays ›
Gene Symbol:
SNAI2
Gene Name:
snail family transcriptional repressor 2
Gene Aliases:
-
Chromosome Location:
Chr.14: 19941414 - 19945016 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03226775_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SNAI2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001034538.2 NP_001029710.1 2 - 3 731 72
GenBank mRNA BC105149.1 - 2 - 3 732 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
osteoblast differentiation
epithelial to mesenchymal transition involved in endocardial cushion formation
cell migration involved in endocardial cushion formation
negative regulation of cell adhesion involved in substrate-bound cell migration
Notch signaling pathway
sensory perception of sound
negative regulation of keratinocyte proliferation
negative regulation of vitamin D biosynthetic process
neural crest cell development
positive regulation of cell migration
negative regulation of chondrocyte differentiation
regulation of chemokine production
positive regulation of histone acetylation
desmosome disassembly
pigmentation
negative regulation of DNA damage response, signal transduction by p53 class mediator
positive regulation of fat cell differentiation
regulation of osteoblast differentiation
white fat cell differentiation
palate development
epithelium development
cartilage morphogenesis
regulation of branching involved in salivary gland morphogenesis
negative regulation of vitamin D receptor signaling pathway
cellular response to epidermal growth factor stimulus
cellular response to ionizing radiation
negative regulation of canonical Wnt signaling pathway
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
negative regulation of stem cell proliferation
regulation of bicellular tight junction assembly
negative regulation of anoikis
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
metal ion binding
E-box binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0