This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03225425_g1
See other PGAM1 GE Assays ›
Gene Symbol:
PGAM1
Gene Name:
phosphoglycerate mutase 1
Gene Aliases:
-
Chromosome Location:
Chr.26: 18649174 - 18655669 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03225425_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PGAM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001034054.1 NP_001029226.1 3 - 4 601 78
GenBank mRNA BC103115.1 - 3 - 4 621 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

mutase

Gene Ontology Categories:

Function(s) Process(es)

glycolytic process
regulation of glycolytic process
regulation of pentose-phosphate shunt
respiratory burst
bisphosphoglycerate mutase activity
phosphoglycerate mutase activity
hydrolase activity
protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0