This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Bt03225368_m1
See other SNCA GE Assays ›
Gene Symbol:
SNCA
Gene Name:
synuclein alpha
Gene Aliases:
-
Chromosome Location:
Chr.6: 34852799 - 35000324 on Build ARS-UCD1.2
Species:
Bovine
Species Specific ID (Flybase ID):
-
Assay ID Bt03225368_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SNCA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001034041.2 NP_001029213.1 2 - 3 177 65
GenBank mRNA BC102402.1 - 2 - 3 179 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

microglial cell activation
positive regulation of neurotransmitter secretion
synaptic transmission, dopaminergic
fatty acid metabolic process
neutral lipid metabolic process
phospholipid metabolic process
mitochondrial membrane organization
adult locomotory behavior
regulation of glutamate secretion
regulation of dopamine secretion
positive regulation of peptidyl-serine phosphorylation
cellular response to oxidative stress
SNARE complex assembly
regulation of locomotion
dopamine biosynthetic process
response to drug
mitochondrial ATP synthesis coupled electron transport
regulation of macrophage activation
negative regulation of neuron apoptotic process
regulation of long-term neuronal synaptic plasticity
synaptic vesicle endocytosis
synaptic vesicle transport
synapse organization
regulation of acyl-CoA biosynthetic process
protein oligomerization
excitatory postsynaptic potential
long-term synaptic potentiation
regulation of reactive oxygen species metabolic process
copper ion binding
histone binding
identical protein binding
arachidonic acid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0